Capillaries for Shimadzu LC systems
Lieferant: Agilent
8001-0818 8001-0821 8001-0822 8001-0824
HEWL8001-0818EA 0 EUR
HEWL8001-0818 HEWL8001-0821 HEWL8001-0822 HEWL8001-0824
Capillaries for Shimadzu LC systems
Schläuche Chromatographie-Schläuche
Stainless steel capillaries and tubing for Shimadzu HPLC Systems. Ideal for most standard applications.
  • Made of ultra-clean stainless steel tubing
  • Ideal for most standard applications, and the best choice for reliable high-pressure sealing
  • Smooth inner surface for the lowest backpressure
  • Precision cutting for burr-free, square-cut ends without inner-diameter distortion
  • Both ends accept standard fittings
  • Predefined lengths for specific path locations to avoid peak broadening
  • Individually tested to ensure reproducibility
Used within combination with the right fittings, they are ideal for LC system plumbing, providing an inert surface, tight, leak-free connections, and zero dead volumes.