Capillaries for waters LC systems
Lieferant: Agilent
8005-0812
HEWL8005-0812EA 0 EUR
HEWL8005-0812
Capillaries for waters LC systems
Schläuche Chromatographie-Schläuche
These capillaries and tubing are made from ultra-clean stainless steel and are suitable for Waters HPLC systems. They are ideal for most standard applications.
  • Reliable high-pressure sealing
  • Smooth inner surface for the lowest backpressure
  • Precision cutting for burr-free, square-cut ends without inner-diameter distortion
  • Both ends accept standard fittings
  • Predefined lengths for specific path locations to avoid peak broadening
  • Individually tested to ensure reproducibility
Used with the right fittings, they are ideal for LC system plumbing, providing an inert surface, tight, leak-free connections, and zero dead volumes.