Connecting capillary, VWD
Lieferant: Agilent
G1314-87302
HEWLG1314-87302EA 81 EUR
HEWLG1314-87302
Connecting capillary, VWD
Schläuche Chromatographie-Schläuche
A comprehensive range of capillaries and fittings are available to protect the integrity of the chromatographic results.
  • Best fit to detector flow cell for optimum chromatographic performance
  • Increased usability without scarifying durability
  • Tight and leak free connection