Avantor® Hichrom PEEKsil™, Tubing
Lieferant: Avantor
HICHHI-1083EA 204 EUR
HICHHI-1083
Avantor® Hichrom PEEKsil™, Tubing
Schläuche Chromatographie-Schläuche
PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica sheathed by PEEK tubing giving it excellent chemical compatibility.