Avantor® Hichrom PEEKsil™, Fused Silica Tubing
Lieferant: Avantor
HICHHI-1097EA 26.5 EUR
HICHHI-1097
Avantor® Hichrom PEEKsil™, Fused Silica Tubing
Schläuche Chromatographie-Schläuche
The maximum operating temperature is 100 °C.
  • pH range 0 to 10
  • Used for micro and nano scale HPLC and capillary electrophorosis
Manufactured from synthetic fused silica with a polyimide coating.