Anti-IgG Goat Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: MP Biomedicals
0855150
ICNA0855150EA 395 EUR
ICNA0855150
Anti-IgG Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
This product is the lyophilized powder of fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat IgG fraction to human IgG Fc and buffer salts,The total protein is measured using the Biuret procedure with bovine albumin as the standard; F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the IgG fraction is standardized by equivalence-point precipitation. The IgG fraction is tested for purity and specificity at 40 mg/mL using immunoelectrophoresis. The product is mostly goat IgG; no trace of albumin is detected.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR