Product is rabbit IgG (whole molecule) covalently coupled to Sepharose-4B, and packed in a polystyrene chromatography column.IgG affinity gel contains a proprietary ligand that retains most protein found in serum, ascites and culture supernatants, while allowing IgG to pass through the support and be collected in the flow-through fraction. Because this gel is not a bind-and-release support, it is extremely fast and gentle to antibodies, resulting in antibody preparations of high purity and high activity. IgG affinity gel works with antibodies from a variety of species and subclasses, many of which do not purify efficiently with Protein A or Protein G. The resulting recovery and purity of the IgG isolated by this method rivals that obtained from the same samples using bind-and-release supports such as Protein A or Protein G. Furthermore, antibodies purified with this gel are recovered ready for use in immunoassays or subsequent conjugation or modification steps; there is no need to neutralize or desalt the recovered antibody to remove harsh elution buffers.
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit