Anti-BSA Rabbit Polyclonal Antibody
Lieferant: MP Biomedicals
08651111
ICNA08651111EA 185 EUR
ICNA08651111
Anti-BSA Rabbit Polyclonal Antibody
Antikörper
The antiserum is in 0,02 M sodium phosphate, 0,14 M sodium chloride, pH 7,3, 0,05% sodium azide, adjusted to standard titer, filtered through a 0,22 μm filter, vialed and lyophilised.
  • Specificity: This antibody shows a single band of precipitation with whole bovine serum and purified BSA, it has not been assayed for interspecies cross-reactivity
  • Can be reconstitute with 2 ml of distilled water
Type: Primary
Antigen: Bovine Serum Albumin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine