Anti-KRT8 Mouse Monoclonal Antibody () [clone: AE5]

Lieferant: MP Biomedicals
08691431
ICNA08691431EA 691 EUR
ICNA08691431
Anti-KRT8 Mouse Monoclonal Antibody () [clone: AE5]
Antikörper
This antibody is specific for the basic 64K keratin which, together with the acidic 55K keratin, has been designated as a marker for "corneal-type differentiation".

The AE5 monoclonal anti-keratin was prepared against human epidermal keratin by hybridoma technique using P3 x 63 Ag8 myeloma cells.

Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: AE5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR