Anti-MLANA Mouse Monoclonal Antibody [clone: A103]

Lieferant: IMMUNOLOGIC
ILM022111-C01 ILM022111-C05 ILM022111-C1
ILM022111-C01EA 87.3 EUR
ILM022111-C01 ILM022111-C05 ILM022111-C1
Anti-MLANA Mouse Monoclonal Antibody [clone: A103]
Antikörper
Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: A103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR