Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]

Lieferant: IMMUNOLOGIC
ILM07211-C01 ILM07211-C05 ILM07211-C1
ILM07211-C01EA 112 EUR
ILM07211-C01 ILM07211-C05 ILM07211-C1
Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Chimpanzee, Common Marmoset
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR