Anti-CEACAM5 Mouse Polyclonal Antibody [clone: CEA31]

Lieferant: IMMUNOLOGIC
ILM1053-C01 ILM1053-C05 ILM1053-C1
ILM1053-C01EA 47.4 EUR
ILM1053-C01 ILM1053-C05 ILM1053-C1
Anti-CEACAM5 Mouse Polyclonal Antibody [clone: CEA31]
Antikörper
Type: Primary
Antigen: CEACAM5
Clonality: Polyclonal
Clone: CEA31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR