Anti-NKX2-1 Mouse Monoclonal Antibody [clone: SPT24]

Lieferant: IMMUNOLOGIC
ILM105846-C01 ILM105846-C05 ILM105846-C1
ILM105846-C01EA 159 EUR
ILM105846-C01 ILM105846-C05 ILM105846-C1
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: SPT24]
Antikörper
Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: SPT24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR