Anti-CHGA Mouse Monoclonal Antibody [clone: LK2H10]

Lieferant: IMMUNOLOGIC
ILM1353-C01 ILM1353-C05 ILM1353-C1
ILM1353-C01EA 55.7 EUR
ILM1353-C01 ILM1353-C05 ILM1353-C1
Anti-CHGA Mouse Monoclonal Antibody [clone: LK2H10]
Antikörper
Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: LK2H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Pig, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR