Anti-NF Mouse Monoclonal Antibody [clone: 2F11]

Lieferant: IMMUNOLOGIC
ILM1503-C01 ILM1503-C05 ILM1503-C1
ILM1503-C01EA 138 EUR
ILM1503-C01 ILM1503-C05 ILM1503-C1
Anti-NF Mouse Monoclonal Antibody [clone: 2F11]
Antikörper
Type: Primary
Antigen: NF
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR