Anti-PGR Rabbit Monoclonal Antibody [clone: Y85]

Lieferant: IMMUNOLOGIC
ILM2073-C01 ILM2073-C05 ILM2073-C1
ILM2073-C01EA 354 EUR
ILM2073-C01 ILM2073-C05 ILM2073-C1
Anti-PGR Rabbit Monoclonal Antibody [clone: Y85]
Antikörper
Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: Y85
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR