Anti-CDX2 Rabbit Monoclonal Antibody [clone: EPR2764Y]

Lieferant: IMMUNOLOGIC
ILM2353-C01 ILM2353-C05 ILM2353-C1
ILM2353-C01EA 197 EUR
ILM2353-C01 ILM2353-C05 ILM2353-C1
Anti-CDX2 Rabbit Monoclonal Antibody [clone: EPR2764Y]
Antikörper
Type: Primary
Antigen: CDX2
Clonality: Monoclonal
Clone: EPR2764Y
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR