Anti-PAX2 Rabbit Monoclonal Antibody [clone: EP3251]

Lieferant: IMMUNOLOGIC
ILM3112-C01 ILM3112-C05 ILM3112-C1
ILM3112-C01EA 292 EUR
ILM3112-C01 ILM3112-C05 ILM3112-C1
Anti-PAX2 Rabbit Monoclonal Antibody [clone: EP3251]
Antikörper
Type: Primary
Antigen: PAX2
Clonality: Monoclonal
Clone: EP3251
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR