Anti-S100B Mouse Polyclonal Antibody [clone: 4C4.9]

Lieferant: IMMUNOLOGIC
ILM3303-C01 ILM3303-C05 ILM3303-C1
ILM3303-C01EA 78.8 EUR
ILM3303-C01 ILM3303-C05 ILM3303-C1
Anti-S100B Mouse Polyclonal Antibody [clone: 4C4.9]
Antikörper
Type: Primary
Antigen: S100B
Clonality: Polyclonal
Clone: 4C4.9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat, Cow
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR