Anti-KRT14 mouse Monoclonal Antibody [clone: LL002]

Lieferant: IMMUNOLOGIC
ILM3861-C01 ILM3861-C05 ILM3861-C1
ILM3861-C01EA 74.9 EUR
ILM3861-C01 ILM3861-C05 ILM3861-C1
Anti-KRT14 mouse Monoclonal Antibody [clone: LL002]
Antikörper
Type: Primary
Antigen: KRT14
Clonality: Monoclonal
Clone: LL002
Conjugation: Unconjugated
Epitope:
Host: mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR