Anti-BAP1 Mouse Monoclonal Antibody [clone: C-4]

Lieferant: IMMUNOLOGIC
ILM3919-C01 ILM3919-C05 ILM3919-C1
ILM3919-C01EA 162 EUR
ILM3919-C01 ILM3919-C05 ILM3919-C1
Anti-BAP1 Mouse Monoclonal Antibody [clone: C-4]
Antikörper
Type: Primary
Antigen: BAP1
Clonality: Monoclonal
Clone: C-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR