Anti-KRT20 Mouse Monoclonal Antibody [clone: KS20.8]

Lieferant: IMMUNOLOGIC
ILM5477-C01 ILM5477-C05 ILM5477-C1
ILM5477-C01EA 215 EUR
ILM5477-C01 ILM5477-C05 ILM5477-C1
Anti-KRT20 Mouse Monoclonal Antibody [clone: KS20.8]
Antikörper
Cytokeratin 20 antibody reacts primarily with gastric and intestinal epithelium, urothelium, and Merkel cells. Anti-Cytokeratin 20 is useful in the differentiation of specific types of simple epithelial cells of the urinary tract as well as normal and malignantly transformed epithelia. Studies have identified the presence of Cytokeratin 20 in adenocarcinomas of the colon, stomach, pancreas and biliary system. Additionally, mucinous ovarian tumors, transitional-cell and merkel cell carcinomas have shown reactivity. In contrast, squamous cell carcinomas and adenocarcinomas of the breast, lung and endometrium, non-mucinous tumors of the ovary, and small cell carcinomas are non-reactive.

  • Visualization: cytoplasmic
  • Control: colon carcinoma
  • Dilution Range: 1:200-1:500

Type: Primary
Antigen: KRT20
Clonality: Monoclonal
Clone: KS20.8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR