Anti-TP53 Mouse Monoclonal Antibody [clone: BP53-12 ]

Lieferant: IMMUNOLOGIC
ILM7157-C01 ILM7157-C05 ILM7157-C1
ILM7157-C01EA 78.8 EUR
ILM7157-C01 ILM7157-C05 ILM7157-C1
Anti-TP53 Mouse Monoclonal Antibody [clone: BP53-12 ]
Antikörper
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: BP53-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Dog, Hamster, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR