Anti-KRT5/KRT6A Mouse Monoclonal Antibody [clone: D5N/16B4]

Lieferant: IMMUNOLOGIC
ILM7773-C01 ILM7773-C05 ILM7773-C1
ILM7773-C01EA 159 EUR
ILM7773-C01 ILM7773-C05 ILM7773-C1
Anti-KRT5/KRT6A Mouse Monoclonal Antibody [clone: D5N/16B4]
Antikörper
Type: Primary
Antigen: KRT5/KRT6A
Clonality: Monoclonal
Clone: D5N/16B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR