Anti-CD5 Mouse Monoclonal Antibody [clone: C5/473]

Lieferant: IMMUNOLOGIC
ILM9229-C01 ILM9229-C05 ILM9229-C1
ILM9229-C01EA 112 EUR
ILM9229-C01 ILM9229-C05 ILM9229-C1
Anti-CD5 Mouse Monoclonal Antibody [clone: C5/473]
Antikörper
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: C5/473
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR