Anti-DNTT Rabbit Polyclonal Antibody

Lieferant: IMMUNOLOGIC
ILP0049-C01 ILP0049-C05 ILP0049-C1
ILP0049-C01EA 260 EUR
ILP0049-C01 ILP0049-C05 ILP0049-C1
Anti-DNTT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Calf, Rat, Mouse, Guinea pig, Cat, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR