Anti-NAPSA Rabbit Polyclonal Antibody

Lieferant: IMMUNOLOGIC
ILP3523-C01 ILP3523-C05 ILP3523-C1
ILP3523-C01EA 171 EUR
ILP3523-C01 ILP3523-C05 VWRKILP3523-C1
Anti-NAPSA Rabbit Polyclonal Antibody
Antikörper
Napsin is a pepsin-like aspartic proteinase, in the A1 clan of the AA clade of proteinases. There are two closely related napsins, napsin A and napsin B. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. It is expressed as a single chain protein in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.1-5

  • Visualization: cytoplasmic
  • Control: lung adenocarcinoma, kidney
  • Dilution Range: 1:100-1:500

References
1. Jagirdar, J. Arch Pathol Lab Med. 2008; 132:384-96.
2. Dejmek A, et al. Diagn Cytopathol. 2007; 35:493-7.
3. Inamura K, et al. Am J Surg Pathol. 2005; 29:660-5.
4. Bishop JA, et al. Hum Pathol. 2010; 41:20-5.
5. Ye J, et al. Appl Immunohistochem Mol Morphol. 2011; 19:313-317.

Type: Primary
Antigen: NAPSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR