Anti-Cathepsin B (Ab-1) (CA10) Mouse Monoclonal Antibody [clone: CA10]

Lieferant: Merck Millipore (Oncogene)
IM27L-100
IM27L-100EA 563 EUR
IM27L-100
Anti-Cathepsin B (Ab-1) (CA10) Mouse Monoclonal Antibody [clone: CA10]
Antikörper
Type: Primary
Antigen: Cathepsin B (Ab-1) (CA10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR