Anti-MMP-7 (Ab-1) (141-7B2) Mouse Monoclonal Antibody
Lieferant: Merck Millipore (Calbiochem‎)
IM40-100UG
IM40-100UGEA 435 EUR
IM40-100UG
Anti-MMP-7 (Ab-1) (141-7B2) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MMP-7 (Ab-1) (141-7B2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human