Anti-MT1-MMP (Ab-4) (113-5B7) Mouse Monoclonal Antibody

Lieferant: Merck Millipore (Oncogene)
IM57-100
IM57-100EA 555 EUR
IM57-100
Anti-MT1-MMP (Ab-4) (113-5B7) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MT1-MMP (Ab-4) (113-5B7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR