Anti-IgG Bovine Polyclonal Antibody (Biotin)

Lieferant: ImmunoReagents
BO-003-CBIO
IMMRBO-003-CBIOEA 103 EUR
IMMRBO-003-CBIO
Anti-IgG Bovine Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Bovine
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR