Anti-IgG Goat Polyclonal Antibody (Biotin)
Lieferant: ImmunoReagents
GT-003-ZBIO
IMMRGT-003-ZBIOEA 103 EUR
IMMRGT-003-ZBIO
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen