Anti-IgG Horse Polyclonal Antibody
Lieferant: ImmunoReagents
HR-003-CBIO
IMMRHR-003-CBIOEA 103 EUR
IMMRHR-003-CBIO
Anti-IgG Horse Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Horse
Isotype:
Reactivity:
Bestellen