Anti-IgG Donkey Polyclonal Antibody (DyLight® 650)

Lieferant: ImmunoReagents
IR1177
IMMRIR1177EA 135 EUR
IMMRIR1177
Anti-IgG Donkey Polyclonal Antibody (DyLight® 650)
Antikörper
The ImmunoReagents' DyLight® 650 conjugates provide vibrant far-red fluorescence with comparable or improved performance compared to other dyes for fluorescent applications. DyLight® 650 dye conjugates have tight excitation/emission spectra that are ideal for multi-color applications. DyLight® 650 conjugated secondary antibodies are available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 652 nm / 672 nm Emission Color = Far Red (Similar Dyes: Alexa Fluor 647, Cy5)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Standards: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Bestellinformation: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR