Anti-IgG Donkey Polyclonal Antibody (DyLight® 550)

Lieferant: ImmunoReagents
IR1194
IMMRIR1194EA 268 EUR
IMMRIR1194
Anti-IgG Donkey Polyclonal Antibody (DyLight® 550)
Antikörper
The ImmunoReagents' DyLight® 550 conjugates provide vibrant orange-to-red fluorescence with better performance than other rhodamine derivatives, including Alexa Fluor 555, TRITC and Cy3 for fluorescent applications. DyLight® 550 conjugates have tight excitation/emission spectra that are ideal for multiColour applications. DyLight® 550 conjugated secondary antibodies are used in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 562 nm/576 nm Emission Colour = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, TRITC).

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Standards: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.

Bestellinformation: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR