Anti-IgG Donkey Polyclonal Antibody (DyLight® 350)

Lieferant: ImmunoReagents
IR1215
IMMRIR1215EA 260 EUR
IMMRIR1215
Anti-IgG Donkey Polyclonal Antibody (DyLight® 350)
Antikörper
The ImmunoReagents' DyLight® 350 conjugates are a derivative of AMCA (Aminomethylcoumarine acetate), which a blue fluorescent dye commonly is used as a contrasting dye for double- and triple-labeling used in a wide array of applications. The UV-excitable DyLight® 350 has spectral properties that are very similar to those displayed by AMCA, including identical excitation wavelengths and a large stokes shift. The emission wavelength of DyLight® 350 has a 10-nm blue shift over AMCA, though, that provides an advantage in multiplexing with green fluorophores. (Excitation/Emission = 353 nm/432 nm Emission Colour = Blue (Similar Dyes: Alexa Fluor 350, AMCA).

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Standards: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.

Bestellinformation: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR