Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: ImmunoReagents
IR1277
IMMRIR1277EA 230 EUR
IMMRIR1277
Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
ImmunoReagents’ FITC Conjugates are used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. Fluorescein is typically excited by the 488 nm line of an argon laser, and emission is collected at 518 nm. FITC antibody conjugates are yellow-orange in Colour but emit a yellow-green Colour upon Em 518 nm. FITC antibody conjugates are sensitive to light. The typical FITC to antibody ratio (F/P) is >4.0 Excitation/Emission = 494 nm/518 nm Emission Colour = Green (Similar Dyes: Alexa Fluor 488, Cy2, DyLight ® 488).

  • Origin: US

Bestellen

TECHNISCHE DATEN

Antigen-Symbol IgG
Antigen-Name Immunoglobulin G
Environmentally Preferable
Antikörper-Typ Sekundär
Klonalität Polyklonal
Konjugation FITC (Fluorescein Isothiocyanate)
Reaktivität Rat
Wirt Donkey
Isotyp IgG
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Immunpräzipitation Yes
ImmunFluoreszenz Yes
Durchflusszytometrie Yes
Antikörperfragment F(ab')2
Immunogen Rat IgG
Format Purified IgG
Form Clear, fluorescent yellow liquid
Reinigung Affinity purified using solid phaseRat IgG
Konzentration 1,50 mg/ml (E 1% at 280 nm = 13,0)
Kreuzadsorption Yes
Lagerungspuffer 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1% (w/v) BSA, Protease/IgG free
Lagertemperatur 2...8 °C
Transporttemperatur 2...8 °C
Validierte Anwendungen Fluoresence ELISA, Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR