ImmunoReagents FITC Conjugates are used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. Fluorescein is typically excited by the 488 nm line of an argon laser, and emission is collected at 518 nm. FITC antibody conjugates are yellow-orange in color but emit a yellow-green color upon Em 518 nm. FITC antibody conjugates are sensitive to light. The typical FITC to antibody ratio (F/P) is >4.0 Excitation/Emission = 494 nm / 518 nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, DyLight ® 488).
ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Standards: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Bestellinformation: Products can by customized based on specific assay requirements: contact your VWR representative for more information.
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human