Anti-IgG Goat Polyclonal Antibody (DyLight® 800)
Lieferant: ImmunoReagents
IR1943
IMMRIR1943EA 343 EUR
IMMRIR1943
Anti-IgG Goat Polyclonal Antibody (DyLight® 800)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse