The ImmunoReagents' DyLight® 633 conjugates fluoresces red with physical properties comparable to other 633 dyes. DyLight® 633 is ideal for multi-colour applications. DyLight® 633 is available as reactive labeling agents and as conjugates of secondary antibodies and biotin-binding proteins for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 638 nm/658 nm Emission Colour = Red (Similar Dyes: Alexa Fluor 633)
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Standards: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Bestellinformation: Products can by customised based on specific assay requirements: contact your VWR representative for more information.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 633
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit