Anti-IgG Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: ImmunoReagents
IR2352
IMMRIR2352EA 133 EUR
IMMRIR2352
Anti-IgG Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
TRITC (Tetramethylrhodamine isothiocyanate) is a popular rhodamine derivative pink in colour but emits a red-orange colour upon Emission at 580 nm. TRITC is frequently paired with FITC in double labeling experiments and conjugated antibodies are often used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. These conjugates are best excited with the 532 nm spectral line of the He-Ne laser and are also recommended as a second colour detection reagent in in situ hybridisation applications. (Excitation/Emission = 555 nm/580 nm Emission Colour = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, DyLight 550)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Standards: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Bestellinformation: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR