Anti-IgG Hamster Polyclonal Antibody (Unconjugated)

Lieferant: ImmunoReagents
IR2388
IMMRIR2388EA 222 EUR
IMMRIR2388
Anti-IgG Hamster Polyclonal Antibody (Unconjugated)
Antikörper
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR