Anti- Llama Polyclonal Antibody (Unconjugated)

Lieferant: ImmunoReagents
IR2401
IMMRIR2401EA 103 EUR
IMMRIR2401
Anti- Llama Polyclonal Antibody (Unconjugated)
Antikörper
Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Llama
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR