Rabbit IgG Isotype Control (Europium)

Lieferant: ImmunoReagents
IR2799
IMMRIR2799EA 1130 EUR
IMMRIR2799
Rabbit IgG Isotype Control (Europium)
Antikörper
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Europium
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR