Anti-IgG Donkey Polyclonal Antibody

Lieferant: ImmunoReagents
IR2884
IMMRIR2884EA 162 EUR
IMMRIR2884
Anti-IgG Donkey Polyclonal Antibody
Antikörper
  

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR