Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Lieferant: ImmunoReagents
IR2957
IMMRIR2957EA 102 EUR
IMMRIR2957
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse