Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Lieferant: ImmunoReagents
RB-003-CBIO
IMMRRB-003-CBIOEA 103 EUR
IMMRRB-003-CBIO
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen