Anti-IgG F(c) Rabbit Polyclonal Antibody (Unconjugated)

Lieferant: ImmunoReagents
RB-004-M
IMMRRB-004-MEA 225 EUR
IMMRRB-004-M
Anti-IgG F(c) Rabbit Polyclonal Antibody (Unconjugated)
Antikörper
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR