Anti-IgG Rabbit Polyclonal Antibody (Unconjugated)

Lieferant: ImmunoReagents
RBXSH-003-D
IMMRRBXSH-003-DEA 148 EUR
IMMRRBXSH-003-D
Anti-IgG Rabbit Polyclonal Antibody (Unconjugated)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR