Saponin permeating solution, 0,5% w/v solution in PBS (10X)

Lieferant: Thermo Fisher Scientific

J63209.AK
J63209.AKEA 108 EUR
J63209.AK
Saponin permeating solution, 0,5% w/v solution in PBS (10X)
Elektrophorese-Reagenzien Western Blotting Reagents

Saponin permeating solution, is used as a cell permeabilising agent.


Physical Form: Liquid


Applications: Cell permeabilizing agent

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR